4-Pentylphenylhydrazine hydrochloride ≥98%

Supplier: Matrix Scientific

012575-1G
101802-894EA 133.85 USD
101802-894
4-Pentylphenylhydrazine hydrochloride ≥98%
4-Pentylphenylhydrazine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₉ClN₂
Melting Pt: >210 °C (dec.)
MDL Number: MFCD06245513
CAS Number: 849021-13-2

Order Now

Specification Test Results

Melting point 210°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR