4-Butylphenylhydrazine hydrochloride ≥98%

Supplier: Matrix Scientific

012573-1G
101802-892EA 62.1 USD
101802-892
4-Butylphenylhydrazine hydrochloride ≥98%
4-Butylphenylhydrazine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₇ClN₂
Melting Pt: >215 °C (dec.)
MDL Number: MFCD00800665
CAS Number: 64287-11-2

Order Now

Specification Test Results

Melting point 215°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR