4-Propylphenylhydrazine hydrochloride ≥98%

Supplier: Matrix Scientific

012563-1G
101802-868EA 103.08 USD
101802-868
4-Propylphenylhydrazine hydrochloride ≥98%
4-Propylphenylhydrazine hydrochloride

Hazard: Irritant

Formula: C₉H₁₅ClN₂
Melting Pt: 239…241 °C
MDL Number: MFCD06245506
CAS Number: 350683-67-9

Order Now

Specification Test Results

Melting point 239-241°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR