3-(3,4-Dimethylphenyl)propionic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(3,4-Dimethylphenyl)propanoic acid

007462-1G
101792-834EA 53.85 USD
101792-834
3-(3,4-Dimethylphenyl)propionic acid ≥98%
3-(3,4-Dimethylphenyl)propionic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Melting Pt: 80…82 °C
MDL Number: MFCD03425686
CAS Number: 25173-76-6

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR