3-[5-(Piperidin-4-yl)[1,2,4]oxadiazol-3-yl]pyridine

Supplier: Matrix Scientific

007357-500MG
101792-542EA 116.92 USD
101792-542
3-[5-(Piperidin-4-yl)[1,2,4]oxadiazol-3-yl]pyridine
3-[5-(Piperidin-4-yl)[1,2,4]oxadiazol-3-yl]pyridine

Hazard: Irritant

Formula: C₁₂H₁₄N₄O
MW: 230.27 g/mol
Melting Pt: 80…83 °C
MDL Number: MFCD03426078
CAS Number: 849925-01-5

Order Now

Specification Test Results

Melting point 80-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR