1-(2,6-Dimethoxybenzyl)-N-methylpiperidine-4-amine

Supplier: Matrix Scientific

007347-500MG
101792-520EA 164.62 USD
101792-520
1-(2,6-Dimethoxybenzyl)-N-methylpiperidine-4-amine
1-(2,6-Dimethoxybenzyl)-N-methylpiperidine-4-amine

Hazard: Irritant

Formula: C₁₅H₂₄N₂O₂
MW: 264.37 g/mol
Melting Pt: 58…60 °C
MDL Number: MFCD01319290
CAS Number: 397245-00-0

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR