1-(4,6-Dimethylpyrimidin-2-yl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

007338-500MG
101792-486EA 53.85 USD
101792-486
1-(4,6-Dimethylpyrimidin-2-yl)piperidine-4-carboxylic acid
1-(4,6-Dimethylpyrimidin-2-yl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₂
Melting Pt: 158…160 °C
MDL Number: MFCD03426079
CAS Number: 797028-97-8

Order Now

Specification Test Results

Melting point 158-160°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR