2-[2-(Aminophenyl)thio]-5-bromopyrimidine

Supplier: Matrix Scientific

007331-500MG
101792-472EA 84.62 USD
101792-472
2-[2-(Aminophenyl)thio]-5-bromopyrimidine
2-[2-(Aminophenyl)thio]-5-bromopyrimidine

Hazard: Irritant

Formula: C₁₀H₈BrN₃S
Melting Pt: 131…133 °C
MDL Number: MFCD03426068
CAS Number: 849021-43-8

Order Now

Specification Test Results

Melting point 131-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR