3,6-Dimethoxypyridazine-4-carboxylic acid

Supplier: Matrix Scientific

007327-500MG
101792-462EA 90.77 USD
101792-462
3,6-Dimethoxypyridazine-4-carboxylic acid
3,6-Dimethoxypyridazine-4-carboxylic acid

Hazard: Irritant

Formula: C₇H₈N₂O₄
Melting Pt: 154…156 °C
MDL Number: MFCD03426073
CAS Number: 89694-24-6

Order Now

Specification Test Results

Melting point 154-156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR