1,2-Dimethyl-1H-indole-3-carbonitrile

Supplier: Matrix Scientific

007316-1G
101792-432EA 112.31 USD
101792-432
1,2-Dimethyl-1H-indole-3-carbonitrile
1,2-Dimethyl-1H-indole-3-carbonitrile

Hazard: Irritant

Formula: C₁₁H₁₀N₂
Melting Pt: 103…105 °C
MDL Number: MFCD00460785
CAS Number: 51072-84-5

Order Now

Specification Test Results

Melting point 103-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR