1,3-Dihydro-1-(piperidin-4-yl)-(2H)-indol-2-one

Supplier: Matrix Scientific

007315-500MG
101792-430EA 70.77 USD
101792-430
1,3-Dihydro-1-(piperidin-4-yl)-(2H)-indol-2-one
1,3-Dihydro-1-(piperidin-4-yl)-(2H)-indol-2-one

Hazard: Irritant

Formula: C₁₃H₁₆N₂O
MW: 216.29 g/mol
Melting Pt: 171…174 °C
MDL Number: MFCD03426085
CAS Number: 16223-25-9

Order Now

Specification Test Results

Melting point 171-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR