2-(3-Chlorophenoxy)malondialdehyde

Supplier: Matrix Scientific

007308-500MG
101792-416EA 92.31 USD
101792-416
2-(3-Chlorophenoxy)malondialdehyde
2-(3-Chlorophenoxy)malondialdehyde

Hazard: Irritant

Formula: C₉H₇ClO₃
Melting Pt: 122…125 °C
MDL Number: MFCD03426060
CAS Number: 849021-39-2

Order Now

Specification Test Results

Melting point 122-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR