2-(4-Chlorophenoxy)malondialdehyde

Supplier: Matrix Scientific

007307-500MG
101792-414EA 113.85 USD
101792-414
2-(4-Chlorophenoxy)malondialdehyde
2-(4-Chlorophenoxy)malondialdehyde

Hazard: Irritant

Formula: C₉H₇ClO₃
MW: 198.61 g/mol
Melting Pt: 116…118 °C
MDL Number: MFCD03426063
CAS Number: 849021-40-5

Order Now

Specification Test Results

Melting point 116-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR