2-(3-Trifluoromethylphenyl)malondialdehyde

Supplier: Matrix Scientific

007305-1G
101792-408EA 141.54 USD
101792-408
2-(3-Trifluoromethylphenyl)malondialdehyde
2-(3-Trifluoromethylphenyl)malondialdehyde

Hazard: Irritant

Formula: C₁₀H₇F₃O₂
MW: 216.16 g/mol
Melting Pt: 99…101 °C
MDL Number: MFCD03424496
CAS Number: 493036-49-0

Order Now

Specification Test Results

Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR