2-Chloro-5-(4-methoxyphenyl)pyrimidine

007303-500MG
101792-404EA 1687.81 USD
101792-404
2-Chloro-5-(4-methoxyphenyl)pyrimidine
2-Chloro-5-(4-methoxyphenyl)pyrimidine

Hazard: Irritant

Formula: C₁₁H₉ClN₂O
Melting Pt: 129 °C
MDL Number: MFCD03426059
CAS Number: 27794-03-2

Order Now

Specification Test Results

Melting point 129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR