2-(4-Fluorophenyl)malondialdehyde ≥98%

Supplier: Matrix Scientific

007281-1G
101792-328EA 100 USD
101792-328
2-(4-Fluorophenyl)malondialdehyde ≥98%
2-(4-Fluorophenyl)malondialdehyde

Hazard: Irritant

Formula: C₉H₇FO₂
MW: 166.15 g/mol
Melting Pt: 165…167 °C
MDL Number: MFCD02261897
CAS Number: 493036-47-8

Order Now

Specification Test Results

Melting point 165-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR