2-(4-Bromophenyl)malondialdehyde

Supplier: Matrix Scientific

007277-1G
101792-318EA 130.77 USD
101792-318
2-(4-Bromophenyl)malondialdehyde
2-(4-Bromophenyl)malondialdehyde

Hazard: Irritant

Formula: C₉H₇BrO₂
MW: 227.06 g/mol
Melting Pt: 157…160 °C
MDL Number: MFCD02261895
CAS Number: 709648-68-0

Order Now

Specification Test Results

Melting point 157-160°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR