4-(4-Chlorophenoxy)phenylhydrazine hydrochloride

Supplier: Matrix Scientific

007272-500MG
101792-306EA 115.38 USD
101792-306
4-(4-Chlorophenoxy)phenylhydrazine hydrochloride
4-(4-Chlorophenoxy)phenylhydrazine hydrochloride

Hazard: Irritant/Irritant-Harmful

Formula: C₁₂H₁₂Cl₂N₂O
Melting Pt: 217…219 °C (dec.)
MDL Number: MFCD03426045
CAS Number: 849021-09-6

Order Now

Specification Test Results

Melting point 217-219°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR