4-(tert-Butoxycarbonylamino)benzylamine

Supplier: Matrix Scientific

Synonyms: 4-Aminobenzylamine, 4-BOC protected

007264-500MG
101792-286EA 115.38 USD
101792-286
4-(tert-Butoxycarbonylamino)benzylamine
4-(tert-Butoxycarbonylamino)benzylamine

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₂
MW: 222.29 g/mol
Melting Pt: 107…109 °C
MDL Number: MFCD02183573
CAS Number: 220298-96-4

Order Now

Specification Test Results

Melting point 107-109°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR