2-[(4-Aminophenyl)thio]-4,6-dimethylpyrimidine

Supplier: Matrix Scientific

Synonyms: 4-[(4,6-Dimethylpyrimidin-2-yl)thio]aniline

006974-1G
101791-708EA 83.08 USD
101791-708
2-[(4-Aminophenyl)thio]-4,6-dimethylpyrimidine
2-[(4-Aminophenyl)thio]-4,6-dimethylpyrimidine

Hazard: Irritant

Formula: C₁₂H₁₃N₃S
MW: 231.32 g/mol
Melting Pt: 176…180 °C
MDL Number: MFCD00203030
CAS Number: 102243-12-9

Order Now

Specification Test Results

Melting point 176-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR