2,3-Dihydro-N,N-dimethyl-5(1H)-indolesulfonamide

Supplier: Matrix Scientific

Synonyms: N,N-Dimethylindoline-5-sulphonamide

006970-1G
101791-700EA 184.62 USD
101791-700
2,3-Dihydro-N,N-dimethyl-5(1H)-indolesulfonamide
2,3-Dihydro-N,N-dimethyl-5(1H)-indolesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂S
MW: 226.3 g/mol
Melting Pt: 109…111 °C
MDL Number: MFCD00214476
CAS Number: 99169-99-0

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR