2,3-Dihydro-5-(methylsulfonyl)-(1H)-indole

Supplier: Matrix Scientific

Synonyms: 5-(Methylsulfonyl)indoline5-(Methylsulphonyl)indoline

006967-1G
101791-692EA 461.54 USD
101791-692
2,3-Dihydro-5-(methylsulfonyl)-(1H)-indole
2,3-Dihydro-5-(methylsulfonyl)-(1H)-indole

Hazard: Irritant

Formula: C₉H₁₁NO₂S
MW: 197.26 g/mol
Melting Pt: 129…131 °C
MDL Number: MFCD01319203
CAS Number: 387350-92-7

Order Now

Specification Test Results

Melting point 129-131°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR