Methyl-5-bromonicotinoylacetate
Supplier: Matrix Scientific
Synonyms:
Methyl 3-(5-bromopyridin-3-yl)-3-oxopropanoate
006947-5G
006947-1G
101791-638EA
63.08
USD
101791-638
101791-640
Methyl-5-bromonicotinoylacetate
Methyl-5-bromonicotinoylacetate
Hazard: Irritant
Formula:
C₉H₈BrNO₃ MW: 258.08 g/mol Melting Pt: 93…94 °C |
MDL Number:
MFCD00216527 CAS Number: 205985-97-3 |
Specification Test Results
Melting point | 93-94°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...