N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-4-aminobenzoic acid

Supplier: Matrix Scientific

006919-1G
101791-562EA 149.23 USD
101791-562
N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-4-aminobenzoic acid
N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-4-aminobenzoic acid

Hazard: Irritant

Formula: C₁₄H₁₅N₃O₂
MW: 257.29 g/mol
Melting Pt: 172…174 °C
MDL Number: MFCD00203268
CAS Number: 387350-50-7

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR