4-(4-Methoxyphenyl)thiophene-2-carboxylic acid

Supplier: Matrix Scientific

006918-500MG
101791-560EA 141.54 USD
101791-560
4-(4-Methoxyphenyl)thiophene-2-carboxylic acid
4-(4-Methoxyphenyl)thiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₀O₃S
Melting Pt: 193…196 °C
MDL Number: MFCD00203285
CAS Number: 82437-74-9

Order Now

Specification Test Results

Melting point 193-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR