6-[2-(Pyrrolidin-1-yl)ethyl]pyridine-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 6-[2-(Pyrrolidin-1-yl)ethyl]nicotinic acid

006913-500MG
101791-550EA 123.08 USD
101791-550
6-[2-(Pyrrolidin-1-yl)ethyl]pyridine-3-carboxylic acid
6-[2-(Pyrrolidin-1-yl)ethyl]pyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
Melting Pt: 198…205 °C (dec.)
MDL Number: MFCD01319352
CAS Number: 387350-41-6

Order Now

Specification Test Results

Melting point 198-205°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR