4-(Methylsulfonyl)aniline hydrochloride

Supplier: Matrix Scientific

006908-100G 006908-25G 006908-5G
101791-528EA 33.35 USD
101791-528 101791-532 101791-530
4-(Methylsulfonyl)aniline hydrochloride
4-(Methylsulphonyl)aniline hydrochloride

Hazard: Irritant

Formula: C₇H₁₀ClNO₂S
Melting Pt: >183 °C (dec.)
MDL Number: MFCD00216490
CAS Number: 177662-76-9

Order Now

Specification Test Results

Melting point 183°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR