2,4-Dimethoxypyrimidine-6-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 2,6-Dimethoxypyrimidine-4-carboxylic acid

006884-1G
101791-454EA 203.08 USD
101791-454
2,4-Dimethoxypyrimidine-6-carboxylic acid
2,4-Dimethoxypyrimidine-6-carboxylic acid

Hazard: Irritant

Formula: C₇H₈N₂O₄
MW: 184.15 g/mol
Melting Pt: 162 °C
MDL Number: MFCD01568105
CAS Number: 59864-30-1

Order Now

Specification Test Results

Melting point 162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR