2,6-Bis(dimethylamino)pyrimidine-4-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 2,4-Bis(dimethylamino)pyrimidine-6-carboxylic acid

006882-500MG
101791-450EA 140 USD
101791-450
2,6-Bis(dimethylamino)pyrimidine-4-carboxylic acid
2,6-Bis(dimethylamino)pyrimidine-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₄N₄O₂
MW: 210.23 g/mol
Melting Pt: 164…168 °C
MDL Number: MFCD01568143
CAS Number: 386715-39-5

Order Now

Specification Test Results

Melting point 164-168°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR