6-(Trifluoromethyl)nicotinic acid hydrazide

Supplier: Matrix Scientific

Synonyms: 2-(Trifluoromethyl)pyridine-5-carboxylic acid hydrazide

006827-1G
101791-242EA 289.23 USD
101791-242
6-(Trifluoromethyl)nicotinic acid hydrazide
6-(Trifluoromethyl)nicotinic acid hydrazide

Hazard: Irritant

Formula: C₇H₆F₅N₃O
Melting Pt: 129…131 °C
MDL Number: MFCD01862651
CAS Number: 386715-32-8

Order Now

Specification Test Results

Melting point 129-131°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR