2-Hydroxy-6-(trifluoromethyl)nicotinamide

Supplier: Matrix Scientific

006822-1G
101791-226EA 158.46 USD
101791-226
2-Hydroxy-6-(trifluoromethyl)nicotinamide
2-Hydroxy-6-(trifluoromethyl)nicotinamide

Hazard: Irritant

Formula: C₇H₅F₃N₂O₂
Melting Pt: 220…223 °C
MDL Number: MFCD00243729
CAS Number: 116548-03-9

Order Now

Specification Test Results

Melting point 220-223°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR