(3,4-Dibenzyloxyphenyl)acetonitrile

Supplier: Matrix Scientific

006698-5G 006698-1G
101790-940EA 115.38 USD
101790-940 101790-942
(3,4-Dibenzyloxyphenyl)acetonitrile
(3,4-Dibenzyloxyphenyl)acetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₂₂H₁₉NO₂
Melting Pt: 80 °C
MDL Number: MFCD00060302
CAS Number: 1699-60-1

Order Now

Specification Test Results

Melting point 80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR