4-Isopropyl-3-nitrobenzotrifluoride ≥97%
Supplier: Matrix Scientific
006623-1G
101790-830EA
103.08
USD
101790-830
4-Isopropyl-3-nitrobenzotrifluoride ≥97%
4-Isopropyl-3-nitrobenzotrifluoride
Hazard: Irritant
Formula:
C₁₀H₁₁F₃N₂O₂ Melting Pt: 103…105 °C |
MDL Number:
MFCD00085098 CAS Number: 175277-90-4 |
Specification Test Results
Melting point | 103-105°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...