4-Benzyloxyphenylacetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(4-(Benzyloxy)phenyl)acetonitrile

006571-1G 006571-5G
101790-778EA 40 USD
101790-778 101790-780
4-Benzyloxyphenylacetonitrile ≥98%
4-Benzyloxyphenylacetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₁₅H₁₃NO
MW: 223.28 g/mol
Melting Pt: 65…67 °C
MDL Number: MFCD00016400
CAS Number: 838-96-0

Order Now

Specification Test Results

Melting point 65-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR