3-Benzyloxyphenylacetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(3-(Benzyloxy)phenyl)acetonitrile

006570-1G 006570-5G
101790-774EA 64.62 USD
101790-774 101790-776
3-Benzyloxyphenylacetonitrile ≥98%
3-Benzyloxyphenylacetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₁₅H₁₃NO
MW: 223.28 g/mol
Melting Pt: 48…49 °C
MDL Number: MFCD00016396
CAS Number: 20967-96-8

Order Now

Specification Test Results

Melting point 48-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR