5-Cyclohexyl-o-anisidine ≥98%
Supplier: Matrix Scientific
006557-5G
006557-1G
101790-740EA
738.46
USD
101790-740
101790-738
5-Cyclohexyl-o-anisidine ≥98%
5-Cyclohexyl-o-anisidine
Hazard: Irritant
Formula:
C₁₃H₁₉NO Melting Pt: 66…68 °C |
MDL Number:
MFCD00270102 CAS Number: 206559-52-6 |
Specification Test Results
Melting point | 66-68°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...