4-Benzyloxy-3-methoxyphenylacetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(4-(Benzyloxy)-3-methoxyphenyl)acetonitrile

006544-5G
101790-698EA 224.25 USD
101790-698
4-Benzyloxy-3-methoxyphenylacetonitrile ≥98%
4-Benzyloxy-3-methoxyphenylacetonitrile

Hazard: Toxic

Formula: C₁₆H₁₅NO₂
MW: 253.30 g/mol
Melting Pt: 68…70 °C
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 68-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR