2,7-Diacetylfluorene ≥98%

Supplier: Matrix Scientific

006405-25G 006405-100G 006405-5G
101790-520EA 56.92 USD
101790-520 101790-522 101790-524
2,7-Diacetylfluorene ≥98%
2,7-Diacetylfluorene

Hazard: Irritant

Formula: C₁₇H₁₄O₂
MW: 250.292 g/mol
Boiling Pt: 459 °C
Melting Pt: 178…179 °C (173 °C)
Density: 1.183
Flash Pt: 171 °C
MDL Number: MFCD00045332
CAS Number: 961-27-3

Order Now

Specification Test Results

Melting point 178-179°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR