Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
3-(Trifluoromethyl)pyrazole-4-carboxylic Acid Ethyl Ester
, Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate
006377-1G
006377-5G
101790-456EA
32.31
USD
101790-456
101790-458
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate ≥97%
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Purity: 97%
CAS: 155377-19-8
MDL Number: MFCD03407995
Molecular Formula: C7H7F3N2O2
Molecular Weight: 208,14
Hazard: Irritant
Formula:
C₇H₇F₃N₂O₂ MW: 208.14 g/mol Melting Pt: 145…148 °C |
MDL Number:
MFCD00052083 CAS Number: 155377-19-8 |
Specification Test Results
Melting point | 145-148°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...