2-(1H)-Tetrahydropyrimidinethione

Supplier: Matrix Scientific

Synonyms: Tetrahydropyrimidine-2(1H)-thione

006359-25G
101790-414EA 56.92 USD
101790-414
2-(1H)-Tetrahydropyrimidinethione
2-(1H)-Tetrahydropyrimidinethione

TCA listed: T
Hazard: Irritant

Formula: C₄H₈N₂S
MW: 116.19 g/mol
Melting Pt: 189 °C
MDL Number: MFCD00060526
CAS Number: 2055-46-1

Order Now

Specification Test Results

Melting point 189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR