5-(Methylthio)-1H-tetrazole ≥97%

Supplier: Matrix Scientific

006297-5G 006297-1G
101790-346EA 41.54 USD
101790-346 101790-348
5-(Methylthio)-1H-tetrazole ≥97%
5-(Methylthio)-1H-tetrazole

Hazard: Irritant

Formula: C₂H₄N₄S
Melting Pt: 150…151 °C (dec.)
MDL Number: MFCD00068726
CAS Number: 29515-99-9

Order Now

Specification Test Results

Melting point 150-151°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR