N-(4-Fluorophenyl)maleamic acid ≥97%

Supplier: Matrix Scientific

006102-5G
101790-086EA 56.92 USD
101790-086
N-(4-Fluorophenyl)maleamic acid ≥97%
N-(4-Fluorophenyl)maleamic acid

Hazard: Irritant

Formula: C₁₀H₈FNO₃
Melting Pt: 209 °C
MDL Number: MFCD00082643
CAS Number: 60252-79-1

Order Now

Specification Test Results

Melting point 209°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR