Tetrafluoroterephthalic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
2,3,5,6-Tetrafluoroterephthalic acid
005941-25G
005941-100G
101789-808EA
201.54
USD
101789-808
101789-806
Tetrafluoroterephthalic acid ≥97%
Tetrafluoroterephthalic acid
Hazard: Irritant
Formula:
C₈H₂F₄O₄ MW: 238.0900 g/mol Melting Pt: 283…284 °C |
MDL Number:
MFCD00012301 CAS Number: 652-36-8 |
Specification Test Results
Melting point | 283-284°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...