2-Phenylbenzonitrile ≥98%
Supplier: Matrix Scientific
Synonyms:
[1,1'-Biphenyl]-2-carbonitrile
005697-5G
005697-1G
101789-378EA
83.08
USD
101789-378
101789-380
2-Phenylbenzonitrile ≥98%
2-Phenylbenzonitrile
Hazard: Irritant
Formula:
C₁₃H₉N MW: 179.2173 g/mol Melting Pt: 35…37 °C |
MDL Number:
MFCD00060669 CAS Number: 24973-49-7 |
Specification Test Results
Melting point | 35-37°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...