3,4-Diethoxybenzhydrazide

Supplier: Matrix Scientific

005654-5G 005654-1G
101789-290EA 40 USD
101789-290 101789-292
3,4-Diethoxybenzhydrazide
3,4-Diethoxybenzhydrazide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃
Melting Pt: 164…166 °C
MDL Number: MFCD01310808
CAS Number: 380482-31-5

Order Now

Specification Test Results

Melting point 164-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR