2-(3-Hydroxy-n-propyl)-5-(trifluoromethyl)benzimidazole

Supplier: Matrix Scientific

005651-1G 005651-5G
101789-280EA 101.54 USD
101789-280 101789-282
2-(3-Hydroxy-n-propyl)-5-(trifluoromethyl)benzimidazole
2-(3-Hydroxy-n-propyl)-5-(trifluoromethyl)benzimidazole

Hazard: Irritant

Formula: C₁₁H₁₁F₃N₂O
MW: 244.22 g/mol
Melting Pt: 139…141 °C
MDL Number: MFCD00067735
CAS Number: 175135-15-6

Order Now

Specification Test Results

Melting point 139-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR