2-Isopropoxy-6-(2,2,2-trifluoroethoxy)benzonitrile ≥95%
Supplier: Matrix Scientific
005650-250MG
005650-1G
101789-276EA
30.77
USD
101789-276
101789-278
2-Isopropoxy-6-(2,2,2-trifluoroethoxy)benzonitrile ≥95%
2-Isopropoxy-6-(2,2,2-trifluoroethoxy)benzonitrile
Purity: 95+%
Formula:
C₁₂H₁₂F₃NO₂ Melting Pt: 88…90 °C |
CAS Number:
175204-05-4 |
Specification Test Results
Melting point | 88-90°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...