4-Trifluoromethylphthalic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)phthalic acid

005435-5G
101788-946EA 538.46 USD
101788-946
4-Trifluoromethylphthalic acid ≥95%
4-Trifluoromethylphthalic acid

Hazard: Irritant

Formula: C₉H₅F₃O₄
MW: 234.13 g/mol
Melting Pt: 151 °C
MDL Number: MFCD00671572
CAS Number: 835-58-5

Order Now

Specification Test Results

Melting point 151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR