4-(Trifluoromethyl)benzoic anhydride

Supplier: Matrix Scientific

005430-5G
101788-936EA 150.77 USD
101788-936
4-(Trifluoromethyl)benzoic anhydride
4-(Trifluoromethyl)benzoic anhydride

Hazard: Irritant

Formula: C₁₆H₈F₆O₃
Melting Pt: 130 °C
MDL Number: MFCD00671577
CAS Number: 25753-16-6

Order Now

Specification Test Results

Melting point 130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR