4-(Trifluoromethoxy)benzaldoxime ≥97%

Supplier: Matrix Scientific

005312-1G
101788-660EA 264.62 USD
101788-660
4-(Trifluoromethoxy)benzaldoxime ≥97%
4-(Trifluoromethoxy)benzaldoxime

Hazard: Irritant

Formula: C₈H₆F₃NO₂
Melting Pt: 41 °C
MDL Number: MFCD00210176
CAS Number: 150162-39-3

Order Now

Specification Test Results

Melting point 41°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR